EMAIL THIS PAGE TO A FRIEND
58118 Sigma-Aldrich

Iodotrimethylsilane

purum, ≥98.0% (AT)

Synonym: TMIS, Trimethyliodosilane

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
InChI Key   CSRZQMIRAZTJOY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
contains   ~1% copper as stabilizer
refractive index   n20/D 1.471(lit.)
bp   106 °C(lit.)
density   1.406 g/mL at 25 °C(lit.)
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Versatile reagent, e.g. for the cleavage of ethers, esters, carbamates and ketals, for the deoxygenation of sulfoxides, for the synthesis of iodides, etc. Reviews; Powerful silylating agent

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2924 3/PG 2
WGK Germany 
3
Flash Point(F) 
-24 °F
Flash Point(C) 
-31 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

195529 Iodotrimethylsilane, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?