EMAIL THIS PAGE TO A FRIEND
58120 Sigma-Aldrich

3-Iodo-L-tyrosine

≥99.0% (HPLC)

Synonym: 3-Monoiodo-L-tyrosine

  • CAS Number 70-78-0

  • Linear Formula IC6H3-4-(OH)CH2CH(NH2)CO2H

  • Molecular Weight 307.09

  •  Beilstein Registry Number 2941266

  •  EC Number 200-744-8

  •  MDL number MFCD00002608

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis, I - Z,
InChI Key   UQTZMGFTRHFAAM-ZETCQYMHSA-N
assay   ≥99.0% (HPLC)
optical activity   [α]20/D −3.1±0.3°, c = 2% in 0.1 M HCl
mp   204-208 °C (dec.)
  210 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Tyrosine hydroxylase inhibitor.

Other Notes

Inhibitor (Ki ~ 500 nM) of tyrosine hydroxylase

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

I8250 3-Iodo-L-tyrosine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?