EMAIL THIS PAGE TO A FRIEND
58150 Sigma-Aldrich

5-Iodouridine

≥97.0% (HPLC)

Synonym: 2,4-Dihydroxy-5-iodo-1-β-D-ribofuranosylpyrimidine

  • CAS Number 1024-99-3

  • Empirical Formula (Hill Notation) C9H11IN2O6

  • Molecular Weight 370.10

  •  Beilstein Registry Number 33665

  •  EC Number 213-833-1

  •  MDL number MFCD00006532

  •  PubChem Substance ID 24881038

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   RKSLVDIXBGWPIS-UAKXSSHOSA-N
assay   ≥97.0% (HPLC)
optical activity   [α]20/D −23.5±1°, c = 1% in H2O
mp   205-207 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

852597 5-Iodouridine, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?