• USA Home
  • 58168 - Ionomycin calcium salt from Streptomyces conglobatus

EMAIL THIS PAGE TO A FRIEND
58168 Sigma-Aldrich

Ionomycin calcium salt from Streptomyces conglobatus

≥80% (HPLC)

  • CAS Number 56092-82-1

  • Empirical Formula (Hill Notation) C41H70CaO9

  • Molecular Weight 747.07

  •  Beilstein Registry Number 1416163

  •  MDL number MFCD00083634

  •  PubChem Substance ID 329758403

Purchase

Description

Other Notes

Nonfluorescent ionophoric antibiotic which exhibits a greater selectivity for Ca2+ over Mg2+ than does A23187

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Biochem/physiol Actions

Ca2+ ionophore that is more effective than A23187 as a mobile ion carrier for Ca2+; non-fluorescent; used to study Ca2+ transport across biological membranes; induces apoptotic degeneration of embryonic cortical neurons.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NO0650000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I0634 Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?