EMAIL THIS PAGE TO A FRIEND
58180 Sigma-Aldrich

β-Ionone

purum, ≥95.0% (GC)

Synonym: 4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one

  • CAS Number 14901-07-6

  • Empirical Formula (Hill Notation) C13H20O

  • Molecular Weight 192.30

  •  Beilstein Registry Number 1909544

  •  EC Number 201-224-3

  •  MDL number MFCD00001549

  •  PubChem Substance ID 329758405

Purchase

Properties

Related Categories Aspalathus linearis (Rooibos tea), Building Blocks, C13 to C14, Carbonyl Compounds, Carthamus tinctorius (Safflower oil),
InChI Key   PSQYTAPXSHCGMF-BQYQJAHWSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.52(lit.)
  n20/D 1.521
bp   126-128 °C/12 mmHg(lit.)
density   0.945 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
EN0500000
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

I12603 β-Ionone, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?