EMAIL THIS PAGE TO A FRIEND
58190 Sigma-Aldrich

Iridium

powder, puriss., 99.9%

Purchase

Properties

InChI Key   GKOZUEZYRPOHIO-UHFFFAOYSA-N
grade   puriss.
assay   99.9%
form   powder
resistivity   4.71 μΩ-cm
bp   4130 °C(lit.)
mp   2450 °C(lit.)
density   22.65 g/cm3(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3089 4.1 / PGII
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?