• USA Home
  • 58219 - ISA (ionic strength adjustment solution: 1 M MgSO4)

EMAIL THIS PAGE TO A FRIEND
58219 Sigma-Aldrich

ISA (ionic strength adjustment solution: 1 M MgSO4)

Synonym: Magnesium sulfate solution

  • CAS Number 7487-88-9

  • Empirical Formula (Hill Notation) MgSO4

  • Molecular Weight 120.37

  •  MDL number MFCD00011110

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionic strength adjustor solutions (ISA) More...
InChI Key   CSNNHWWHGAXBCP-UHFFFAOYSA-L

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Solution used to establish a constant ionic strength in a sample solution for determinations with ISE

Same product — same quality — now Sigma-Aldrich brand

Preparation Note

prepared with high purity salts

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?