• USA Home
  • 58225 - Isethionic acid ammonium salt

EMAIL THIS PAGE TO A FRIEND
58225 Sigma-Aldrich

Isethionic acid ammonium salt

purum

Synonym: 2-Hydroxyethanesulfonic acid ammonium salt

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acid Salts, Sulfur Compounds More...
InChI Key   LLOHIFXFHGMBNO-UHFFFAOYSA-N
grade   purum
mp   138-141 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
KI7750000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

268445 Isethionic acid ammonium salt, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?