EMAIL THIS PAGE TO A FRIEND
58480 Sigma-Aldrich

Isobutylamine

purum, ≥98.0% (GC)

Synonym: 1-Amino-2-methylpropane

  • CAS Number 78-81-9

  • Linear Formula (CH3)2CHCH2NH2

  • Molecular Weight 73.14

  •  Beilstein Registry Number 385626

  •  EC Number 201-145-4

  •  MDL number MFCD00008146

  •  PubChem Substance ID 329758417

Purchase

Properties

InChI Key   KDSNLYIMUZNERS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
impurities   ≤1% water
refractive index   n20/D 1.397(lit.)
  n20/D 1.397
bp   64-71 °C(lit.)
mp   −85 °C(lit.)
density   0.736 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1214 - class 3 - PG 2 - Isobutylamine
WGK Germany 
3
RTECS 
NP9900000
Flash Point(F) 
15.8 °F
Flash Point(C) 
-9 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?