• USA Home
  • 58752 - Isoquinoline-1-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
58752 Sigma-Aldrich

Isoquinoline-1-carboxylic acid

technical, ≥97.0% (T)

  • CAS Number 486-73-7

  • Empirical Formula (Hill Notation) C10H7NO2

  • Molecular Weight 173.17

  •  Beilstein Registry Number 129177

  •  EC Number 207-639-6

  •  MDL number MFCD00006901

  •  PubChem Substance ID 329758453

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Isoquinolines
InChI Key   XAAKCCMYRKZRAK-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (T)
mp   164 °C (dec.)(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?