• USA Home
  • 58775 - Isocitric Dehydrogenase (NADP) from porcine heart

EMAIL THIS PAGE TO A FRIEND
58775 Sigma-Aldrich

Isocitric Dehydrogenase (NADP) from porcine heart

3-8 U/mg protein (~12 mg/ml)

Synonym: threo-Ds-Isocitrate: NADP+ oxidoreductase, ICDH, IDH

Purchase

Description

Other Notes

Sales restrictions may apply

Unit Definition

1 U corresponds to the amount of enzyme which catalyzes the conversion of 1 μmol isocitrate to α-ketoglutarate at pH 7.4 and 37°C

Physical form

50% glycerol; EDTA buffer salts, pH ~6.0

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?