• USA Home
  • 58810 - Triethoxy(3-isocyanatopropyl)silane

EMAIL THIS PAGE TO A FRIEND
58810 Sigma-Aldrich

Triethoxy(3-isocyanatopropyl)silane

purum, ≥95.0% (GC)

Synonym: (3-Isocyanatopropyl)triethoxysilane

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Trialkoxysilanes
InChI Key   FRGPKMWIYVTFIQ-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.42(lit.)
bp   283 °C(lit.)
density   0.999 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3390 8(6.1) / PGI
WGK Germany 
3
RTECS 
VV6691000
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

413364 3-(Triethoxysilyl)propyl isocyanate, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?