• USA Home
  • 58822 - Ethyl isocyanoacetate

EMAIL THIS PAGE TO A FRIEND
58822 Sigma-Aldrich

Ethyl isocyanoacetate

purum, ≥98.0% (GC)

  • CAS Number 2999-46-4

  • Linear Formula CNCH2COOCH2CH3

  • Molecular Weight 113.11

  •  Beilstein Registry Number 3588613

  •  EC Number 221-077-9

  •  MDL number MFCD00000007

  •  PubChem Substance ID 24881207

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanides, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   FPULFENIJDPZBX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.418(lit.)
  n20/D 1.418
bp   194-196 °C(lit.)
density   1.035 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Caution

discolors to brown on storage

Other Notes

Reviews; Synthesis of α-amino acids

This product has been replaced by 226319-ALDRICH | Ethyl isocyanoacetate 95%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
183.2 °F
Flash Point(C) 
84 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

226319 Ethyl isocyanoacetate, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?