EMAIL THIS PAGE TO A FRIEND
58930 Sigma-Aldrich

Isonicotinic acid

purum, ≥99.0% (T)

Synonym: 4-Picolinic acid, Pyridine-4-carboxylic acid

  • CAS Number 55-22-1

  • Empirical Formula (Hill Notation) C6H5NO2

  • Molecular Weight 123.11

  •  Beilstein Registry Number 109599

  •  EC Number 200-228-2

  •  MDL number MFCD00006429

  •  PubChem Substance ID 24881074

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   TWBYWOBDOCUKOW-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   ≥300 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
NS1103000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?