EMAIL THIS PAGE TO A FRIEND
58950 Sigma-Aldrich

Ethyl isonicotinate

purum, ≥99.0% (GC)

  • CAS Number 1570-45-2

  • Empirical Formula (Hill Notation) C8H9NO2

  • Molecular Weight 151.16

  •  Beilstein Registry Number 122942

  •  EC Number 216-379-2

  •  MDL number MFCD00006428

  •  PubChem Substance ID 329758534

Purchase

Properties

Related Categories Building Blocks, C8, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   MCRPKBUFXAKDKI-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.501(lit.)
  n20/D 1.501
bp   92 °C/8 mmHg(lit.)
solubility   H2O: soluble
  ethanol: soluble
  methanol: soluble
density   1.009 g/mL at 25 °C(lit.)
fluorescence   λex 360 nm; λem 450 nm in H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NA 1993 / PGIII
WGK Germany 
3
RTECS 
NS1450000
Flash Point(F) 
190.4 °F
Flash Point(C) 
88 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

104736 Ethyl isonicotinate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?