Linear Formula H2O
EC Number 231-791-2
MDL number MFCD00011332
Popular Documents: Specification Sheet (PDF)
| Related Categories | Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, Blends - CHROMASOLV for HPLC, Blends- CHROMASOLV for HPLC, |
| contains | 0.05 % (v/v) trifluoroacetic acid |
18 L in VerSA-Flow™
4×4 L in glass bottle
Product filtered through a 0.2 μm filter
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?