EMAIL THIS PAGE TO A FRIEND
59015 Sigma-Aldrich

Isononylamine

purum, mixture of isomeric nonylamines, ≥97.0% (GC)

  • Empirical Formula (Hill Notation) C9H21N

  • Molecular Weight 143.27

  •  Beilstein Registry Number 2069449

  •  EC Number 248-653-2

Purchase

Properties

grade   purum
assay   ≥97.0% (GC)
  ~90% 3,5,5-trimethylhexylamine basis
density   0.786 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGII
WGK Germany 
2
Flash Point(F) 
132.8 °F
Flash Point(C) 
56 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?