• USA Home
  • 590754 - Acetonitrile solution

EMAIL THIS PAGE TO A FRIEND
590754 Sigma-Aldrich

Acetonitrile solution

contains 0.1 % (v/v) acetic acid

  • CAS Number 75-05-8

  • Linear Formula CH3CN

  • Molecular Weight 41.05

  •  Beilstein Registry Number 4124158

  •  EC Number 200-835-2

  •  MDL number MFCD00001878

  •  PubChem Substance ID 57651668

Purchase

Description

Preparation Note

Product filtered through a 0.2 μm filter

Price and Availability


Durapore Membrane Filters

Fluoropore and Omnipore Membrane Filters
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
41 °F
Flash Point(C) 
5 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?