EMAIL THIS PAGE TO A FRIEND
59080 Sigma-Aldrich

2-Methylbutane

purum, ≥95.0% (GC)

Synonym: Isopentane

  • CAS Number 78-78-4

  • Linear Formula CH3CH2CH(CH3)2

  • Molecular Weight 72.15

  •  Beilstein Registry Number 1730723

  •  EC Number 201-142-8

  •  MDL number MFCD00009338

  •  PubChem Substance ID 24881299

Purchase

Properties

vapor density   2.6 (vs air)
vapor pressure   11.17 psi ( 20 °C)
InChI Key   QWTDNUCVQCZILF-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
autoignition temp.   788 °F
expl. lim.   8.3 %
impurities   ~2% pentane
refractive index   n20/D 1.354(lit.)
  n20/D 1.354
bp   30 °C(lit.)
density   0.62 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 1 - Pentanes
WGK Germany 
2
RTECS 
EK4430000
Flash Point(F) 
-59.8 °F
Flash Point(C) 
-51 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?