EMAIL THIS PAGE TO A FRIEND
59102 Sigma-Aldrich

Isopentylamine

purum, ≥98.0% (GC)

Synonym: 1-Amino-3-methylbutane, Isoamylamine

  • CAS Number 107-85-7

  • Linear Formula (CH3)2CHCH2CH2NH2

  • Molecular Weight 87.16

  •  Beilstein Registry Number 1209230

  •  EC Number 203-526-0

  •  MDL number MFCD00008203

  •  PubChem Substance ID 329758579

Purchase

Properties

InChI Key   BMFVGAAISNGQNM-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.408(lit.)
  n20/D 1.408
bp   95-97 °C(lit.)
density   0.751 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1106 8(3) / PGII
WGK Germany 
1
Flash Point(F) 
30.2 °F
Flash Point(C) 
-1 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

126810 Isopentylamine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?