EMAIL THIS PAGE TO A FRIEND
59160 Sigma-Aldrich

Isopentyl nitrite

≥97.0% (GC)

Synonym: Isoamyl nitrite

  • CAS Number 110-46-3

  • Linear Formula (CH3)2CHCH2CH2ONO

  • Molecular Weight 117.15

  •  Beilstein Registry Number 969510

  •  MDL number MFCD00002057

  •  PubChem Substance ID 329758595

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Organic Nitrates/Nitrites More...
InChI Key   OWFXIOWLTKNBAP-UHFFFAOYSA-N
assay   ≥97.0% (GC)
contains   ~2% sodium carbonate as stabilizer
refractive index   n20/D 1.386(lit.)
  n20/D 1.387
bp   99 °C(lit.)
density   0.872 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1113 - class 3 - PG 2 - Amyl nitrites
WGK Germany 
1
RTECS 
NT0187500
Flash Point(F) 
-4 °F
Flash Point(C) 
-20 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

150495 Isopentyl nitrite, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?