• USA Home
  • 59220 - Dimethyl isophthalate

EMAIL THIS PAGE TO A FRIEND
59220 Sigma-Aldrich

Dimethyl isophthalate

purum, ≥98.0% (GC)

  • CAS Number 1459-93-4

  • Linear Formula C6H4-1,3-(CO2CH3)2

  • Molecular Weight 194.18

  •  Beilstein Registry Number 1912251

  •  EC Number 215-951-9

  •  MDL number MFCD00008433

Purchase

Properties

vapor density   6.8 (vs air)
vapor pressure   93.7 mmHg ( 208 °C)
InChI Key   VNGOYPQMJFJDLV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   64-68 °C(lit.)
  64-68 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
NT2540000
Flash Point(F) 
343.4 °F
Flash Point(C) 
173 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

194239 Dimethyl isophthalate, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?