• USA Home
  • 59595 - N-Isopropylmethylamine

EMAIL THIS PAGE TO A FRIEND
59595 Sigma-Aldrich

N-Isopropylmethylamine

purum, ≥98.0% (GC)

Synonym: N-Methylisopropylamine

  • CAS Number 4747-21-1

  • Linear Formula (CH3)2CHNHCH3

  • Molecular Weight 73.14

  •  Beilstein Registry Number 1730877

  •  EC Number 225-266-7

  •  MDL number MFCD00042859

  •  PubChem Substance ID 24881606

Purchase

Properties

vapor pressure   4.15 psi ( 20 °C)
InChI Key   XHFGWHUWQXTGAT-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.384(lit.)
  n20/D 1.384
bp   50-53 °C(lit.)
density   0.702 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGI
WGK Germany 
3
Flash Point(F) 
-25.6 °F
Flash Point(C) 
-32 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?