• USA Home
  • 59688 - HiCrome Listeria Selective Supplement

EMAIL THIS PAGE TO A FRIEND
59688 Sigma-Aldrich

HiCrome Listeria Selective Supplement

for microbiology

Purchase

Properties

Related Categories Media, Microbiology, Supplements More...
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
storage temp.   2-8°C

Description

Components

(One vial is sufficient for 500 mL medium)
Ceftazidime 2.0 mg, Amphotericin B 2.5 mg

General description

A selective supplement recommended for rapid and direct identification of Listeria species.

Legal Information

HiCrome is a trademark of HiMedia Laboratories Pvt. Ltd.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?