• USA Home
  • 59740 - Isopropyl β-D-1-thiogalactopyranoside

EMAIL THIS PAGE TO A FRIEND
59740 Sigma-Aldrich

Isopropyl β-D-1-thiogalactopyranoside

≥99.0% (TLC)

Synonym: IPTG, Isopropyl β-D-thiogalactoside

  • CAS Number 367-93-1

  • Empirical Formula (Hill Notation) C9H18O5S

  • Molecular Weight 238.30

  •  Beilstein Registry Number 4631

  •  EC Number 206-703-0

  •  MDL number MFCD00063273

  •  PubChem Substance ID 329758709

Purchase

Properties

description   non-ionic
InChI Key   BPHPUYQFMNQIOC-NXRLNHOXSA-N
assay   ≥99.0% (TLC)
optical activity   [α]20/D −32.0±1.0°, c = 1% in H2O (dry matter)
ign. residue   ≤0.2%
loss   ≤6% loss on drying
storage temp.   −20°C

Description

Application

IPTG is commonly used in cloning procedures that require induction of β-galactosidase activity. It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli. IPTG functions by binding to the lacI repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ.

Other Notes

Non-metabolizable galactose analog.

IPTG is an inducer of β-galactosidase activity in bacteria and is used to detect lac gene activity during cloning experiments

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I5502 Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?