EMAIL THIS PAGE TO A FRIEND
59862 Sigma-Aldrich

Ethyl isovalerate

purum, ≥97.0% (GC)

  • CAS Number 108-64-5

  • Linear Formula (CH3)2CHCH2COOC2H5

  • Molecular Weight 130.18

  •  Beilstein Registry Number 1744677

  •  EC Number 203-602-3

  •  MDL number MFCD00009203

  •  PubChem Substance ID 329758631

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor pressure   7.5 mmHg ( 20 °C)
InChI Key   PPXUHEORWJQRHJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.396(lit.)
  n20/D 1.398
bp   131-133 °C(lit.)
mp   −99 °C(lit.)
density   0.864 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
2
RTECS 
NY1504000
Flash Point(F) 
80.6 °F
Flash Point(C) 
27 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?