EMAIL THIS PAGE TO A FRIEND
59890 Sigma-Aldrich

Isovaleryl chloride

purum, ≥98.0% (AT)

Synonym: 3-Methylbutyryl chloride

  • CAS Number 108-12-3

  • Linear Formula (CH3)2CHCH2COCl

  • Molecular Weight 120.58

  •  Beilstein Registry Number 741910

  •  EC Number 203-552-2

  •  MDL number MFCD00000738

  •  PubChem Substance ID 329758633

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   ISULZYQDGYXDFW-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
refractive index   n20/D 1.416(lit.)
  n20/D 1.416
bp   115-117 °C(lit.)
density   0.982 g/mL at 20 °C
  0.989 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
87.8 °F
Flash Point(C) 
31 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

157422 Isovaleryl chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?