EMAIL THIS PAGE TO A FRIEND
59945 Sigma-Aldrich

Isoxanthopterin

≥97.5% (HPLC)

Synonym: 2-Amino-4,7-dihydroxy-1,3,5,8-tetraazanaphthalene, 2-Amino-4,7-dihydroxypteridine, 2-Amino-4,7-pteridinediol

  • CAS Number 529-69-1

  • Empirical Formula (Hill Notation) C6H5N5O2

  • Molecular Weight 179.14

  •  Beilstein Registry Number 181594

  •  EC Number 208-469-5

  •  MDL number MFCD00006696

  •  PubChem Substance ID 329758638

Purchase

Properties

InChI Key   GLKCOBIIZKYKFN-UHFFFAOYSA-N
assay   ≥97.5% (HPLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?