EMAIL THIS PAGE TO A FRIEND
60065 Sigma-Aldrich

Potassium benzoate

ReagentPlus®, ≥99.0%

Synonym: Benzoic acid potassium salt

  • CAS Number 582-25-2

  • Linear Formula C6H5COOK

  • Molecular Weight 160.21

  •  Beilstein Registry Number 3719165

  •  EC Number 209-481-3

  •  MDL number MFCD00013061

  •  PubChem Substance ID 24881786

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Potassium Salts, Research Essentials,
grade   ReagentPlus®
InChI Key   XAEFZNCEHLXOMS-UHFFFAOYSA-M
assay   ≥99.0%

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?