• USA Home
  • 60232 - Potassium Phosphate Monobasic solution

EMAIL THIS PAGE TO A FRIEND
60232 Sigma-Aldrich

Potassium Phosphate Monobasic solution

for HPLC, concentrate II, ampule

Synonym: Monopotassium phosphate, Potassium dihydrogen phosphate solution

Purchase

Properties

grade   for HPLC
InChI Key   GNSKLFRGEWLPPA-UHFFFAOYSA-M
quality   ampule
λ   in concentrate
UV absorption   λ: 210 nm Amax: 0.1
  λ: 240 nm Amax: 0.06
  λ: 260 nm Amax: 0.04
  λ: 300 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for RP gradient test
  corresponds to standard for filter test

Description

Packaging

concentrate (~0.66 M) in ampules for 6x1 l 0.01 M ready-to-use solution

Reconstitution

recommendation for use in ion pair chromatography

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?