• USA Home
  • 60242 - Potassium fluoride dihydrate

EMAIL THIS PAGE TO A FRIEND
60242 Sigma-Aldrich

Potassium fluoride dihydrate

spray dried, ReagentPlus®, ≥99.0%

Purchase

Properties

Related Categories Chemical Synthesis, Deprotecting Reagents, Essential Chemicals, Fluoride Sources, Potassium Salts,
grade   ReagentPlus®
InChI Key   MZFXHSQFQLKKFD-UHFFFAOYSA-M
assay   ≥99.0%
mp   41 °C(lit.)

Description

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1812 6.1 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?