• USA Home
  • 60260 - Potassium hexachloroplatinate(IV)

EMAIL THIS PAGE TO A FRIEND
60260 Sigma-Aldrich

Potassium hexachloroplatinate(IV)

puriss., ~40% Pt basis

Synonym: Dipotassium hexachloroplatinate, Dipotassium platinum hexachloride, Platinic potassium chloride, Platinum potassium chloride, Potassium chloroplatinate, Potassium platinum(IV) chloride, Potassium platinum(IV) hexachloride

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3290 6.1/PG 2
WGK Germany 
1
RTECS 
TP1650000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

520861 Potassium hexachloroplatinate(IV), ≥99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?