• USA Home
  • 60340 - Potassium bicarbonate

EMAIL THIS PAGE TO A FRIEND
60340 Sigma-Aldrich

Potassium bicarbonate

purum p.a., ≥99.0% (T), powder, crystals or chunks

Synonym: Potassium hydrogen carbonate

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Potassium, Potassium Salts,
InChI Key   TYJJADVDDVDEDZ-UHFFFAOYSA-M
grade   purum p.a.
assay   ≥99.0% (T)
form   powder, crystals or chunks
solubility   water: soluble(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤20 mg/kg
  Na: ≤200 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?