• USA Home
  • 60344 - Potassium hydrogen difluoride

EMAIL THIS PAGE TO A FRIEND
60344 Sigma-Aldrich

Potassium hydrogen difluoride

purum p.a., ≥99.0% (F)

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Fluorination, Fluorination Reagents, Potassium Salts,
grade   purum p.a.
assay   ≥99.0% (F)
anion traces   chloride (Cl-): ≤100 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  Na: ≤5000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Other Notes

Fluorinating agent; preparation of 2-fluoro-sugars; Monofluorination of inorganic phosphate

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1811 8(6.1) / PGII
WGK Germany 
3
RTECS 
TS6650000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?