• USA Home
  • 60396 - Potassium ionophore IV

EMAIL THIS PAGE TO A FRIEND
60396 Sigma-Aldrich

Potassium ionophore IV

Selectophore, function tested

Synonym: 4-tert-Butyl-2,2,14,14-tetrahomo-2a,14a,dioxacalix[4]arene-tetraacetic acid tetra-tert-butyl ester

  • CAS Number 135463-06-8

  • Empirical Formula (Hill Notation) C70H100O14

  • Molecular Weight 1165.53

  •  MDL number MFCD01868123

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
quality   function tested

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Ionophore used in potassium-selective electrodes

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

NIST2203 Potassium fluoride, NIST SRM 2203, standard for ion-selective electrodes
99373 Potassium ionophore I - cocktail B, Selectophore
70385 2,3-Naphtho-15-crown-5, Selectophore, ≥99.0%
99311 Potassium ionophore I - cocktail A, Selectophore
60397 Potassium ionophore III, Selectophore, function tested
60401 Potassium ionophore II, Selectophore, function tested
60403 Potassium ionophore I, Selectophore, function tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?