• USA Home
  • 60398 - Potassium ionophore I - cocktail B

EMAIL THIS PAGE TO A FRIEND
60398 Sigma-Aldrich

Potassium ionophore I - cocktail B

Selectophore

  •  Beilstein Registry Number 78657

  •  MDL number MFCD00145149

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophore Cocktails More...
grade   Selectophore
InChI Key   FCFNRCROJUBPLU-DNDCDFAISA-N

Description

Components

Composition:
Potassium Ionophore (60403) 5.00 wt %
1,2-Dimethyl-3-nitrobenzene (40870) 93.0 wt %
Potassium tetrakis(4-chlorophenyl)borate (60591) 2.00 wt%

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3172 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?