• USA Home
  • 60426 - Potassium oxalate monohydrate

EMAIL THIS PAGE TO A FRIEND
60426 Sigma-Aldrich

Potassium oxalate monohydrate

puriss. p.a., ACS reagent, ≥99.5% (RT)

Synonym: Ethanedioic acid, Oxalic acid potassium salt

  • CAS Number 6487-48-5

  • Linear Formula (COOK)2 · H2O

  • Molecular Weight 184.23

  •  Beilstein Registry Number 3752576

  •  EC Number 209-506-8

  •  MDL number MFCD00150033

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Biochemicals and Reagents, Chelators,
grade   ACS reagent
InChI Key   QCPTVXCMROGZOL-UHFFFAOYSA-L
grade   puriss. p.a.
assay   ≥99.5% (RT)
impurities   ≤0.002% ammonium (NH4)
pH   7.0-8.5 (25 °C, 50 mg/mL in H2O)
anion traces   chloride (Cl-): ≤10 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤200 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




REDI-DRI
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?