• USA Home
  • 60519 - Potassium thiocyanate

EMAIL THIS PAGE TO A FRIEND
60519 Sigma-Aldrich

Potassium thiocyanate

puriss. p.a., ACS reagent, ≥99.0% (AT)

Synonym: Potassium rhodanide

  • CAS Number 333-20-0

  • Linear Formula KSCN

  • Molecular Weight 97.18

  •  Beilstein Registry Number 3594799

  •  EC Number 206-370-1

  •  MDL number MFCD00011413

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Essential Chemicals, O-P, Puriss p.a. ACS,
grade   ACS reagent
InChI Key   ZNNZYHKDIALBAK-UHFFFAOYSA-M
grade   puriss. p.a.
assay   ≥99.0% (AT)
pH   5.3-8.7 (25 °C, 50 mg/mL in H2O)
mp   173 °C(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤20 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤2 mg/kg
  Fe: ≤1 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  NH4+: ≤30 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤2 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
XL1925000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?