• USA Home
  • 60525 - Potassium-selective liquid ion-exchanger microelectrode cocktail A

EMAIL THIS PAGE TO A FRIEND
60525 Sigma-Aldrich

Potassium-selective liquid ion-exchanger microelectrode cocktail A

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophore Cocktails More...
grade   Selectophore

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Cocktail for microelectrodes for the determination of potassium. Electrodes have a short response time and a low resistance

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?