• USA Home
  • 60586 - Potassium tetrakis(4-tert-butylphenyl)borate

EMAIL THIS PAGE TO A FRIEND
60586 Sigma-Aldrich

Potassium tetrakis(4-tert-butylphenyl)borate

Selectophore, ≥97.0% (C)

Synonym: Tetrakis(4-tert-butylphenyl)boron potassium

Purchase

Properties

Related Categories Additives for ISE, Analytical/Chromatography, Ion Sensor Materials More...
grade   Selectophore
InChI Key   YSPXNGBLXJXCEH-UHFFFAOYSA-N
assay   ≥97.0% (C)
mp   ≥300 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Tetrakis(4-tert-butylphenyl)borate has a very stable structure

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?