EMAIL THIS PAGE TO A FRIEND
60611 Sigma-Aldrich

Camomile oil boiled

Synonym: Chamomile oil boiled

Purchase

Properties

Related Categories Alphabetical Listings, C-D, Flavors and Fragrances More...
refractive index   n20/D 1.47
Organoleptic   floral

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
FL7181000
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?