EMAIL THIS PAGE TO A FRIEND
60748 Sigma-Aldrich

Silica gel 90 CN

for column chromatography, 0.015-0.035 mm

Purchase

Properties

Related Categories Adsorbents (Inorganic), Analytical/Chromatography, LPLC Separation Media, LPLC/TLC/Paper Chromatography, Silica More...
grade   for column chromatography
impurities   ≥0.9% nitrogen
  ≥5.5% carbon
particle size   0.015-0.035 mm

Description

Analysis Note

surface coverage with (−CCH2)3CN

Application

for direct- and reversed phase chromatography

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?