EMAIL THIS PAGE TO A FRIEND
60751 Sigma-Aldrich

Silica gel

high-purity grade, pore size 60 Å, ≥400 mesh particle size

Purchase

Properties

Related Categories Adsorbents (Inorganic), Analytical/Chromatography, Irregular Silica Gel, High-Purity Grade, LPLC Separation Media, LPLC/TLC/Paper Chromatography,
grade   high-purity grade
description   for preparative liquid chromatography
InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
form   powder
mfr. no.   Sigma-Aldrich®
activity acc. to Brockmann and Schodder   2-3
particle size   ≤40 μm
  ≥400 mesh
pore size   0.8 cm3/g pore volume
  60 Å
surface area   500 m2/g
bp   2230 °C
mp   >1600 °C

Description

Application

Silica gel may be used in separation of single-walled carbon nanotube (SWNT) by simple filtration.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?