• USA Home
  • 60754 - C18-Reversed phase silica gel

EMAIL THIS PAGE TO A FRIEND
60754 Sigma-Aldrich

C18-Reversed phase silica gel

for column chromatography, fully endcapped

Synonym: Reversed phase silica gel C18

Purchase

Properties

Related Categories Analytical/Chromatography, LPLC Separation Media, LPLC/TLC/Paper Chromatography, Reversed Phase
grade   for column chromatography
quality   fully endcapped
surface coverage   max. surface coverage (C) ≥15% ±4 μmol/m2
matrix active group   C18 phase
particle size   15-35 μm (≥400 mesh ASTM)
pore size   90 Å pore size
surface area   380 m2/g

Description

Application

for reversed phase and ion pair chromatography

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

60756 C18-Reversed phase silica gel, for column chromatography, fully end-capped

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?