• USA Home
  • 60755 - Silica gel 100 C8-Reversed phase

EMAIL THIS PAGE TO A FRIEND
60755 Sigma-Aldrich

Silica gel 100 C8-Reversed phase

for column chromatography, fully endcapped

Synonym: Reversed phase silica gel 100 C8

Purchase

Properties

Related Categories Analytical/Chromatography, LPLC Separation Media, LPLC/TLC/Paper Chromatography, Reversed Phase
grade   for column chromatography
quality   fully endcapped
surface coverage   surface coverage (C) 10‑12% ±4.0 μmol/m2
particle size   0.015-0.035 mm (≥400 mesh ASTM)

Description

Application

for reversed phase and ion pair chromatography

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?