EMAIL THIS PAGE TO A FRIEND
60775 Sigma-Aldrich

Silica gel

high-purity grade, HF254, without calcium sulfate, with fluorescent indicator, for thin layer chromatography

Purchase

Properties

Related Categories Adsorbents for TLC, Analytical/Chromatography, LPLC/TLC/Paper Chromatography, Silica TLC Adsorbent, with fluorescent indicator, no binder, Silica TLC Adsorbents,
grade   for thin layer chromatography
  high-purity grade
InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
quality   without calcium sulfate, with fluorescent indicator
bp   2230 °C
mp   >1600 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?