• USA Home
  • 60790 - Cobalt(II) acetate tetrahydrate

EMAIL THIS PAGE TO A FRIEND
60790 Sigma-Aldrich

Cobalt(II) acetate tetrahydrate

purum p.a., crystallized, ≥99.0% (KT)

Synonym: Cobaltous acetate tetrahydrate

  • CAS Number 6147-53-1

  • Linear Formula (CH3COO)2Co · 4H2O

  • Molecular Weight 249.08

  •  Beilstein Registry Number 3731397

  •  EC Number 200-755-8

  •  MDL number MFCD00149569

  •  PubChem Substance ID 329759222

Purchase

Properties

Related Categories Cobalt Salts, Essential Chemicals, Research Essentials, Salts
InChI Key   ZBYYWKJVSFHYJL-UHFFFAOYSA-L
grade   purum p.a.
assay   ≥99.0% (KT)
quality   crystallized
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤1000 mg/kg
  Pb: ≤10 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
AG3325000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?