EMAIL THIS PAGE TO A FRIEND
60890 Sigma-Aldrich

Kojic acid

purum, ≥98.0% (HPLC)

Synonym: 2-Hydroxymethyl-5-hydroxy-γ-pyrone, 5-Hydroxy-2-hydroxymethyl-4H-4-pyranone

  • CAS Number 501-30-4

  • Empirical Formula (Hill Notation) C6H6O4

  • Molecular Weight 142.11

  •  Beilstein Registry Number 120895

  •  EC Number 207-922-4

  •  MDL number MFCD00006580

  •  PubChem Substance ID 24882002

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrans
InChI Key   BEJNERDRQOWKJM-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   152-155 °C(lit.)

Description

Biochem/physiol Actions

Tyrosinase inhibitor.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UQ0875000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

K3125 Kojic acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?