EMAIL THIS PAGE TO A FRIEND
60985 Sigma-Aldrich

KRANEP Agar

for microbiology

Synonym: Potassium thiocyanate Actidione Sodium azide Egg-yolk Pyruvate Agar Basis

Purchase

Properties

Related Categories A - M, All Food Control Media (alphabetical), All Solid Media/Agars, Media, Media by Application,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   actidione, 0.041 g/L
  agar, 13.5 g/L
  disodium hydrogen phosphate, 2 g/L
  lithium chloride, 5.1 g/L
  D(-)-mannitol, 5.1 g/L
  meat peptone, 10 g/L
  potassium thiocyanate, 25.5 g/L
  sodium azide, 0.05 g/L
  sodium chloride, 3 g/L
  sodium pyruvate, 8.2 g/L
final pH   6.9±0.2 (25 °C)
suitability   Micrococcus
  selective for Staphylococcus

Description

Application

Selective agar for the isolation of all staphylococci and micrococci from food according to Sinell and Baumgart.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?