61163 Sigma-Aldrich

Copper(I) bromide

purum p.a., ≥98.0% (RT)

DOWNLOAD MSDS (PDF)

Synonym: Cuprous bromide

  • CAS Number 7787-70-4

  • Linear Formula CuBr

  • Molecular Weight 143.45

  •  EC Number 232-131-6

  •  MDL number MFCD00010969

  •  PubChem Substance ID 24882063

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Copper More...
grade   purum p.a.
assay   ≥98.0% (RT)
impurities   ≤3% Cu(II)Br2
mp   504 °C(lit.)
density   4.71 g/mL at 25 °C(lit.)
anion traces   sulfate (SO42-): ≤5000 mg/kg
cation traces   Ca: ≤500 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Fe: ≤500 mg/kg
  K: ≤100 mg/kg
  Na: ≤5000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤500 mg/kg
  Zn: ≤100 mg/kg

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?